siRNA Details
Gene Symbol: DRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001388.4 | NP_001379.1 | 13 | 1977 | ||
GenBank mRNA | X80754.1 | - | 13 | 1867 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DRG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001388.4 | NP_001379.1 | 13 | 1977 | ||
GenBank mRNA | X80754.1 | - | 13 | 1867 |