siRNA Details
| Gene Symbol: EIF4B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300821.1 | NP_001287750.1 | 5 | 725 | ||
| NM_001417.5 | NP_001408.2 | 5 | 725 | |||
| GenBank mRNA | AB076839.1 | - | 4 | 499 | ||
| AK293731.1 | - | 5 | 527 | |||
| AK299329.1 | - | 5 | 527 | |||
| AK299526.1 | - | 4 | 411 | |||
| AK310244.1 | - | 6 | 664 | |||
| BC073139.1 | - | 5 | 515 | 6527540 | ||
| BC073154.1 | - | 5 | 515 | 6015183 | ||
| BC098437.1 | - | 5 | 509 | 6459393 | ||
| BP236621.1 | - | 5 | 544 | |||
| CR456958.1 | - | 4 | 499 | |||

EIF4B
Genomic Map