siRNA Details
| Gene Symbol: EMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001424.5 | NP_001415.1 | 5 | 731 | ||
| XM_006720864.3 | XP_006720927.1 | 5 | 725 | |||
| GenBank mRNA | BC009687.1 | - | 5 | 622 | 3895094 | |
| U52100.1 | - | 4 | 566 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMP2| Gene Symbol: EMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001424.5 | NP_001415.1 | 5 | 731 | ||
| XM_006720864.3 | XP_006720927.1 | 5 | 725 | |||
| GenBank mRNA | BC009687.1 | - | 5 | 622 | 3895094 | |
| U52100.1 | - | 4 | 566 | |||