siRNA Details
| Gene Symbol: GPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001448.2 | NP_001439.2 | 9 | 1937 | ||
| GenBank mRNA | AF030186.1 | - | 9 | 1937 | ||
| AF064826.1 | - | 9 | 1842 | |||
| AK304207.1 | - | 9 | 1662 | |||
| AY052833.1 | - | 9 | 1861 | |||
| AY358507.1 | - | 9 | 2223 | |||
| BC017166.1 | - | 9 | 2231 | 3922774 | ||

GPC4
Genomic Map