siRNA Details
| Gene Symbol: CAMK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323374.1 | NP_001310303.1 | 10 | 843 | ||
| NM_001323375.1 | NP_001310304.1 | 10 | 875 | |||
| NM_001323376.1 | NP_001310305.1 | 7 | 845 | |||
| NM_001323377.1 | NP_001310306.1 | 8 | 603 | |||
| NM_001744.5 | NP_001735.1 | 9 | 991 | |||
| GenBank mRNA | AK311662.1 | - | 7 | 700 | ||
| AK312840.1 | - | 9 | 846 | |||
| BC016695.1 | - | 9 | 835 | |||
| BC025687.1 | - | 9 | 833 | 5205478 | ||
| D30742.1 | - | 9 | 821 | |||
| L17000.1 | - | 9 | 785 | |||
| L24959.1 | - | 10 | 843 | |||

CAMK4
Genomic Map