siRNA Details
| Gene Symbol: CD22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185099.1 | NP_001172028.1 | 13 | 2794 | ||
| NM_001185100.1 | NP_001172029.1 | 13 | 2939 | |||
| NM_001185101.1 | NP_001172030.1 | 12 | 2527 | |||
| NM_001278417.1 | NP_001265346.1 | 13 | 2886 | |||
| NM_001771.3 | NP_001762.2 | 14 | 3058 | |||
| GenBank mRNA | AK225625.1 | - | 13 | 2776 | ||
| AK225694.1 | - | 14 | 3040 | |||
| AK301177.1 | - | 14 | 2921 | |||
| BQ054599.1 | - | 1 | 450 | |||
| X59350.1 | - | 14 | 3018 | |||

CD22
Genomic Map