siRNA Details
| Gene Symbol: CD58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144822.1 | NP_001138294.1 | 3 | 539 | ||
| NM_001779.2 | NP_001770.1 | 3 | 539 | |||
| NR_026665.1 | 3 | 539 | ||||
| XM_017002869.1 | XP_016858358.1 | 3 | 539 | |||
| GenBank mRNA | AK291980.1 | - | 3 | 472 | ||
| BC005930.1 | - | 3 | 485 | 4285827 | ||
| BT019816.1 | - | 3 | 418 | |||
| BT019817.1 | - | 3 | 418 | |||
| CR456939.1 | - | 3 | 418 | |||
| X06296.1 | - | 3 | 430 | |||
| Y00636.1 | - | 3 | 427 | |||

CD58
Genomic Map