siRNA Details
| Gene Symbol: CHN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025201.3 | NP_001020372.2 | 6 | 530 | ||
| NM_001822.5 | NP_001813.1 | 5 | 529 | |||
| NR_038133.1 | 5 | 343 | ||||
| GenBank mRNA | AB209788.1 | - | 5 | 328 | ||
| AK289941.1 | - | 5 | 459 | |||
| AK300890.1 | - | 6 | 524 | |||
| BC011393.1 | - | 5 | 501 | 3871858 | ||
| BQ893027.1 | - | 6 | 563 | |||
| DA516095.1 | - | 5 | 539 | |||
| Z22641.1 | - | 5 | 198 | |||

CHN1
Genomic Map