siRNA Details
| Gene Symbol: CCR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164680.1 | NP_001158152.1 | 3 | 1579 | ||
| NM_001837.3 | NP_001828.1 | 3 | 1598 | |||
| NM_178328.1 | NP_847898.1 | 3 | 1588 | |||
| NM_178329.2 | NP_847899.1 | 2 | 1519 | |||
| XM_006712960.3 | XP_006713023.1 | 3 | 1625 | |||
| XM_011533335.2 | XP_011531637.1 | 3 | 1705 | |||
| XM_017005685.1 | XP_016861174.1 | 3 | 2508 | |||
| XM_017005686.1 | XP_016861175.1 | 4 | 2689 | |||
| GenBank mRNA | BC110297.1 | - | 2 | 1428 | 5212541 | |

CCR3
Genomic Map