siRNA Details
| Gene Symbol: CCR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301714.1 | NP_001288643.1 | 2 | 1370 | ||
| NM_001301716.1 | NP_001288645.1 | 3 | 1670 | |||
| NM_001301717.1 | NP_001288646.1 | 3 | 1426 | |||
| NM_001301718.1 | NP_001288647.1 | 3 | 1516 | |||
| NM_001838.3 | NP_001829.1 | 3 | 1420 | |||
| GenBank mRNA | AK127810.1 | - | 3 | 1408 | ||
| AK310277.1 | - | 3 | 1670 | |||
| BC035343.1 | - | 3 | 1386 | 5180758 | ||
| L08176.1 | - | 3 | 1410 | |||
| L31581.1 | - | 3 | 1409 | |||

CCR7
Genomic Map