siRNA Details
Gene Symbol: CNR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001841.2 | NP_001832.1 | 2 | 1735 | ||
XM_011540629.2 | XP_011538931.1 | 2 | 2018 | |||
XM_017000261.1 | XP_016855750.1 | 5 | 2710 | |||
GenBank mRNA | EU517121.1 | - | 3 | 1867 | ||
X74328.1 | - | 2 | 1736 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
