siRNA Details
| Gene Symbol: COL19A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001858.5 | NP_001849.2 | 6 | 575 | ||
| XM_011535437.2 | XP_011533739.1 | 6 | 842 | |||
| XM_011535438.2 | XP_011533740.1 | 6 | 570 | |||
| XM_017010253.1 | XP_016865742.1 | 6 | 842 | |||
| XM_017010254.1 | XP_016865743.1 | 6 | 842 | |||
| XM_017010255.1 | XP_016865744.1 | 6 | 781 | |||
| XM_017010256.1 | XP_016865745.1 | 6 | 575 | |||
| XM_017010257.1 | XP_016865746.1 | 6 | 842 | |||
| XM_017010258.1 | XP_016865747.1 | 6 | 575 | |||
| XM_017010259.1 | XP_016865748.1 | 6 | 841 | |||
| GenBank mRNA | AK310621.1 | - | 6 | 565 | ||
| BC113362.1 | - | 6 | 543 | 8322414 | ||
| BC113364.1 | - | 6 | 543 | 8322416 | ||
| D38163.1 | - | 6 | 575 | |||
| L12347.1 | - | 2 | 143 | |||
| M63597.1 | - | 5 | 384 | |||

COL19A1
Genomic Map