siRNA Details
| Gene Symbol: CSNK2A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001896.2 | NP_001887.1 | 12 | 1384 | ||
| XM_005255800.3 | XP_005255857.1 | 10 | 1469 | |||
| XM_005255801.3 | XP_005255858.1 | 12 | 1380 | |||
| XM_017022945.1 | XP_016878434.1 | 8 | 1293 | |||
| GenBank mRNA | BC008812.2 | - | 12 | 1258 | 3951914 | |
| M55268.1 | - | 13 | 1405 | |||

CSNK2A2
Genomic Map