siRNA Details
| Gene Symbol: CYB561 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017916.1 | NP_001017916.1 | 6 | 1497 | ||
| NM_001017917.1 | NP_001017917.1 | 6 | 1330 | |||
| NM_001915.3 | NP_001906.3 | 6 | 1327 | |||
| XM_005257091.1 | XP_005257148.1 | 6 | 1272 | |||
| XM_017024260.1 | XP_016879749.1 | 6 | 1369 | |||
| XM_017024261.1 | XP_016879750.1 | 5 | 1236 | |||
| XM_017024262.1 | XP_016879751.1 | 6 | 1562 | |||
| GenBank mRNA | AK095244.1 | - | 6 | 1544 | ||
| AK301541.1 | - | 5 | 1099 | |||
| BC002976.1 | - | 6 | 1284 | 3535771 | ||
| BC091485.1 | - | 6 | 1298 | 6188259 | ||

CYB561
Genomic Map