siRNA Details
| Gene Symbol: CLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001294338.1 | NP_001281267.1 | 3 | 564 | ||
| NM_001294339.1 | NP_001281268.1 | 3 | 564 | |||
| NM_003993.3 | NP_003984.2 | 3 | 564 | |||
| XM_005244878.1 | XP_005244935.1 | 3 | 546 | |||
| XM_011509143.1 | XP_011507445.1 | 3 | 546 | |||
| XM_011509144.1 | XP_011507446.1 | 3 | 549 | |||
| XM_011509147.2 | XP_011507449.1 | 3 | 536 | |||
| XM_017000241.1 | XP_016855730.1 | 3 | 539 | |||
| XM_017000242.1 | XP_016855731.1 | 3 | 539 | |||
| XM_017000243.1 | XP_016855732.1 | 3 | 539 | |||
| XM_017000244.1 | XP_016855733.1 | 3 | 536 | |||
| GenBank mRNA | AK291995.1 | - | 3 | 408 | ||
| AK303696.1 | - | 4 | 343 | |||
| BC014067.2 | - | 3 | 322 | 3837777 | ||
| BC053603.1 | - | 3 | 482 | 6007375 | ||
| BI562223.1 | - | 3 | 529 | |||
| L29216.1 | - | 3 | 364 | |||
| L29218.1 | - | 3 | 364 | |||

CLK2
Genomic Map