siRNA Details
| Gene Symbol: CDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317227.1 | NP_001304156.1 | 2 | 617 | ||
| NM_001317228.1 | NP_001304157.1 | 3 | 732 | |||
| NM_004061.4 | NP_004052.2 | 5 | 1246 | |||
| XM_011513927.2 | XP_011512229.1 | 3 | 523 | |||
| XM_017008920.1 | XP_016864409.1 | 4 | 873 | |||
| XM_017008921.1 | XP_016864410.1 | 5 | 966 | |||
| GenBank mRNA | AK295123.1 | - | 3 | 732 | ||
| AK314800.1 | - | 2 | 617 | |||
| BC047608.1 | - | 5 | 951 | 5751948 | ||
| L33477.1 | - | 5 | 1247 | |||
| L34057.1 | - | 1 | 252 | |||

CDH12
Genomic Map