siRNA Details
| Gene Symbol: CHN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293069.1 | NP_001279998.1 | 7 | 597 | ||
| NM_001293070.1 | NP_001279999.1 | 7 | 811 | |||
| NM_001293071.1 | NP_001280000.1 | 5 | 716 | |||
| NM_001293072.1 | NP_001280001.1 | 6 | 529 | |||
| NM_004067.3 | NP_004058.1 | 6 | 772 | |||
| XM_011515105.2 | XP_011513407.1 | 9 | 652 | |||
| XM_011515106.2 | XP_011513408.1 | 8 | 613 | |||
| XM_011515107.2 | XP_011513409.1 | 8 | 1395 | |||
| XM_017011721.1 | XP_016867210.1 | 10 | 670 | |||
| XM_017011722.1 | XP_016867211.1 | 9 | 1413 | |||
| GenBank mRNA | AK124021.1 | - | 2 | 1880 | ||
| AK293959.1 | - | 7 | 391 | |||
| AK294032.1 | - | 6 | 529 | |||
| AK294271.1 | - | 7 | 598 | |||
| AK313021.1 | - | 6 | 546 | |||
| BC112155.1 | - | 6 | 517 | 8327623 | ||
| EU732760.1 | - | 5 | 279 | |||
| EU732766.1 | - | 6 | 335 | |||
| GQ924106.1 | - | 7 | 598 | |||
| U28926.1 | - | 6 | 569 | |||

CHN2
Genomic Map