siRNA Details
Gene Symbol: CMKLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142343.1 | NP_001135815.1 | 4 | 1908 | ||
NM_001142344.1 | NP_001135816.1 | 3 | 1695 | |||
NM_001142345.1 | NP_001135817.1 | 3 | 1522 | |||
NM_004072.2 | NP_004063.1 | 3 | 1832 | |||
XM_017018820.1 | XP_016874309.1 | 2 | 1650 | |||
GenBank mRNA | U79527.1 | - | 3 | 1838 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
