siRNA Details
| Gene Symbol: COCH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135058.1 | NP_001128530.1 | 7 | 902 | ||
| NM_004086.2 | NP_004077.1 | 8 | 578 | |||
| XM_017021071.1 | XP_016876560.1 | 8 | 812 | |||
| XM_017021072.1 | XP_016876561.1 | 7 | 951 | |||
| GenBank mRNA | AF006740.1 | - | 8 | 554 | ||
| AK123362.1 | - | 6 | 708 | |||
| AK130273.1 | - | 1 | 27 | |||
| AK292724.1 | - | 8 | 554 | |||
| AY358900.1 | - | 7 | 893 | |||
| BC007230.1 | - | 7 | 865 | 2824361 | ||
| BC018827.2 | - | 8 | 541 | |||

COCH
Genomic Map