siRNA Details
| Gene Symbol: STOM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270526.1 | NP_001257455.1 | 7 | 1307 | ||
| NM_004099.5 | NP_004090.4 | 7 | 1398 | |||
| NM_198194.2 | NP_937837.1 | 3 | 903 | |||
| NR_073037.1 | 8 | 1528 | ||||
| GenBank mRNA | AK304449.1 | - | 8 | 1472 | ||
| AK304518.1 | - | 6 | 1039 | |||
| AK307999.1 | - | 7 | 1341 | |||
| BC010703.1 | - | 7 | 1320 | 3873494 | ||
| BM925356.1 | - | 1 | 480 | |||
| M81635.1 | - | 7 | 1302 | |||

STOM
Genomic Map