siRNA Details
Gene Symbol: EPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004447.5 | NP_004438.3 | 3, 4 | 571 | ||
XM_011520605.2 | XP_011518907.1 | 4, 5 | 631 | |||
GenBank mRNA | AK291777.1 | - | 3, 4 | 376 | ||
AK292931.1 | - | 3, 4 | 228 | |||
AK304860.1 | - | 3, 4 | 571 | |||
BC030010.2 | - | 3, 4 | 383 | 4820933 | ||
BI667617.1 | - | 446 | ||||
U12535.1 | - | 3, 4 | 343 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_007945, NM_001271588, NM_001271595, NM_001271587, AK155022, and others.
Panther Classification:
Gene Ontology Categories:
