siRNA Details
| Gene Symbol: ERH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004450.2 | NP_004441.1 | 4 | 382 | ||
| GenBank mRNA | AK130850.1 | - | 4 | 368 | ||
| BC014301.2 | - | 4 | 365 | 3507241 | ||
| BC071709.1 | - | 4 | 329 | 5503738 | ||
| D85758.1 | - | 4 | 385 | |||
| U66871.1 | - | 4 | 382 | |||

ERH
Genomic Map