siRNA Details
| Gene Symbol: CD81 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297649.1 | NP_001284578.1 | 7 | 726 | ||
| NM_004356.3 | NP_004347.1 | 7 | 844 | |||
| XM_011520492.2 | XP_011518794.1 | 7 | 1078 | |||
| XM_017018598.1 | XP_016874087.1 | 7 | 587 | |||
| GenBank mRNA | AB209380.1 | - | 2 | 4516 | ||
| BC002978.1 | - | 7 | 651 | 3510634 | ||
| BC093047.1 | - | 7 | 842 | 30398483 | ||
| BG397506.1 | - | 8 | 714 | |||
| BT019507.1 | - | 7 | 611 | |||
| BT019508.1 | - | 7 | 611 | |||
| M33680.1 | - | 7 | 849 | |||

CD81
Genomic Map