siRNA Details
| Gene Symbol: CLGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130675.1 | NP_001124147.1 | 6, 7 | 632 | ||
| NM_004362.2 | NP_004353.1 | 5, 6 | 548 | |||
| GenBank mRNA | AK093096.1 | - | 5, 6 | 1111 | ||
| AK290819.1 | - | 5, 6 | 545 | |||
| AK299243.1 | - | 4, 5 | 337 | |||
| AL045602.1 | - | 6, 7 | 576 | |||
| BC028357.2 | - | 5, 6 | 502 | 4830135 | ||
| D86322.1 | - | 5, 6 | 515 | |||

CLGN
Genomic Map