siRNA Details
| Gene Symbol: CEACAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291484.2 | NP_001278413.1 | 8 | 2092 | ||
| NM_001308398.1 | NP_001295327.1 | 8 | 2089 | |||
| NM_004363.5 | NP_004354.3 | 8 | 2092 | |||
| XM_011526322.1 | XP_011524624.1 | 6 | 1527 | |||
| XM_017026145.1 | XP_016881634.1 | 8 | 2058 | |||
| XM_017026146.1 | XP_016881635.1 | 7 | 1806 | |||
| GenBank mRNA | AB852566.1 | - | 6 | 1411 | ||
| AB852567.1 | - | 5 | 877 | |||
| AK223101.1 | - | 8 | 2028 | |||
| BC034671.1 | - | 8 | 2040 | 5184800 | ||
| CR749337.1 | - | 6 | 1134 | |||
| M15042.1 | - | 8 | 1934 | |||
| M16234.1 | - | 4 | 956 | |||
| M29540.1 | - | 8 | 2059 | |||
| X16455.1 | - | 8 | 1931 | |||

CEACAM5
Genomic Map