siRNA Details
| Gene Symbol: CCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004367.5 | NP_004358.2 | 3 | 2206 | ||
| NM_031409.3 | NP_113597.2 | 3 | 2469 | |||
| GenBank mRNA | AK223494.1 | - | 3 | 2125 | ||
| U68030.1 | - | 4 | 2165 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCR6| Gene Symbol: CCR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004367.5 | NP_004358.2 | 3 | 2206 | ||
| NM_031409.3 | NP_113597.2 | 3 | 2469 | |||
| GenBank mRNA | AK223494.1 | - | 3 | 2125 | ||
| U68030.1 | - | 4 | 2165 | |||