siRNA Details
| Gene Symbol: CNN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
| RefSeq | NM_001303499.1 | NP_001290428.1 | 3, 4 | 647 | ||
| NM_001303501.1 | NP_001290430.1 | 3, 4 | 647 | |||
| NM_004368.3 | NP_004359.1 | 3, 4 | 647 | |||
| NM_201277.2 | NP_958434.1 | 3, 4 | 647 | |||
| GenBank mRNA | AK057960.1 | - | 3, 4 | 293 | ||
| AK126391.1 | - | 3, 4 | 312 | |||
| AK222924.1 | - | 3, 4 | 283 | |||
| AK293171.1 | - | 3, 4 | 293 | |||
| AK295271.1 | - | 3, 4 | 263 | |||
| AK297774.1 | - | 4, 5 | 232 | |||
| AK300788.1 | - | 3, 4 | 282 | |||
| AY927514.1 | - | LIT3524 AY927514 |
3, 4 | 231 | ||
| BC141818.1 | - | 3, 4 | 290 | 40148129 | ||
| BC141833.1 | - | 3, 4 | 290 | 40148834 | ||
| BC148264.1 | - | 3, 4 | 282 | 8860204 | ||
| BQ876824.1 | - | 3, 4 | 222 | |||
| BQ919304.1 | - | 3, 4 | 623 | |||
| CR541810.1 | - | 3, 4 | 260 | |||
| CR541831.1 | - | 3, 4 | 260 | |||
| D83735.1 | - | 3, 4 | 287 | |||
| D86059.1 | - | 3, 4 | 263 | |||

CNN2
Genomic Map