siRNA Details
| Gene Symbol: DAG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165928.3 | NP_001159400.2 | 6 | 1407 | ||
| NM_001177634.2 | NP_001171105.1 | 6 | 1350 | |||
| NM_001177635.2 | NP_001171106.1 | 4 | 939 | |||
| NM_001177636.2 | NP_001171107.1 | 5 | 1328 | |||
| NM_001177637.2 | NP_001171108.1 | 4 | 1166 | |||
| NM_001177638.2 | NP_001171109.1 | 4 | 1197 | |||
| NM_001177639.2 | NP_001171110.1 | 3 | 887 | |||
| NM_001177640.2 | NP_001171111.1 | 4 | 1103 | |||
| NM_001177641.2 | NP_001171112.1 | 4 | 1193 | |||
| NM_001177642.2 | NP_001171113.1 | 4 | 1098 | |||
| NM_001177643.2 | NP_001171114.1 | 3 | 1097 | |||
| NM_001177644.2 | NP_001171115.1 | 3 | 1186 | |||
| NM_004393.5 | NP_004384.4 | 3 | 1046 | |||
| GenBank mRNA | AK291692.1 | - | 3 | 888 | ||
| AK297802.1 | - | 1 | 202 | |||
| BC012740.2 | - | 3 | 981 | 4099382 | ||
| BC014616.2 | - | 3 | 981 | 2964740 | ||
| L19711.1 | - | 3 | 1022 | |||

DAG1
Genomic Map