siRNA Details
| Gene Symbol: DFNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127453.1 | NP_001120925.1 | 4 | 548 | ||
| NM_001127454.1 | NP_001120926.1 | 3 | 563 | |||
| NM_004403.2 | NP_004394.1 | 4 | 793 | |||
| XM_017011802.1 | XP_016867291.1 | 3 | 319 | |||
| GenBank mRNA | AF007790.2 | - | 3 | 448 | ||
| AF073308.1 | - | 4 | 517 | |||
| AK094714.1 | - | 6 | 1121 | |||
| AK290775.1 | - | 4 | 524 | |||
| AK314402.1 | - | 4 | 533 | |||
| BC099911.1 | - | 4 | 509 | 5287362 | ||
| BC125065.1 | - | 4 | 515 | 40117291 | ||
| BC125066.1 | - | 3 | 285 | |||
| DA820184.1 | - | 4 | 548 | |||

DFNA5
Genomic Map