siRNA Details
| Gene Symbol: EPHA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288629.1 | NP_001275558.1 | 2, 3 | 403 | ||
| NM_001288630.1 | NP_001275559.1 | 2, 3 | 403 | |||
| NM_004440.3 | NP_004431.1 | 2, 3 | 403 | |||
| XM_005248669.2 | XP_005248726.1 | 2, 3 | 460 | |||
| XM_005248671.2 | XP_005248728.1 | 2, 3 | 460 | |||
| XM_017010365.1 | XP_016865854.1 | 2, 3 | 460 | |||
| XM_017010366.1 | XP_016865855.1 | 2, 3 | 460 | |||
| GenBank mRNA | AB209269.1 | - | 2, 3 | 390 | ||
| AK313529.1 | - | 2, 3 | 375 | |||
| BC027940.1 | - | 2, 3 | 368 | |||
| BC126125.1 | - | 2, 3 | 277 | 8991841 | ||
| BC126151.1 | - | 2, 3 | 277 | 8991867 | ||
| BC143857.1 | - | 2, 3 | 277 | 9052373 | ||
| BC143858.1 | - | 2, 3 | 277 | 9052374 | ||
| DA503795.1 | - | 2, 3 | 403 | |||
| L36642.1 | - | 2, 3 | 347 | |||

EPHA7
Genomic Map