siRNA Details
| Gene Symbol: EPYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004950.4 | NP_004941.2 | 4 | 535 | ||
| XM_011538008.1 | XP_011536310.1 | 3 | 351 | |||
| GenBank mRNA | AK290642.1 | - | 4 | 492 | ||
| BC030958.1 | - | 4 | 535 | 4604494 | ||
| U59111.1 | - | 4 | 488 | |||

EPYC
Genomic Map