siRNA Details
| Gene Symbol: EPYC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004950.4 | NP_004941.2 | 7 | 977 | ||
| XM_011538008.1 | XP_011536310.1 | 6 | 793 | |||
| GenBank mRNA | AK290642.1 | - | 7 | 934 | ||
| BC030958.1 | - | 7 | 977 | 4604494 | ||
| U59111.1 | - | 7 | 930 | |||

EPYC
Genomic Map