siRNA Details
| Gene Symbol: GPR183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004951.4 | NP_004942.1 | 2 | 248 | ||
| XM_017020405.1 | XP_016875894.1 | 2 | 191 | |||
| GenBank mRNA | AK223390.1 | - | 2 | 152 | ||
| AK292091.1 | - | 2 | 172 | |||
| AK313443.1 | - | 2 | 248 | |||
| AW402166.1 | - | 2 | 200 | |||
| BC020752.1 | - | 2 | 152 | 4722289 | ||
| L08177.1 | - | 1 | 107 | |||
More Information
Set Membership: |
|

GPR183
Genomic Map