siRNA Details
| Gene Symbol: FOXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085471.1 | NP_001078940.1 | 7 | 1831 | ||
| NM_005197.3 | NP_005188.2 | 6 | 1882 | |||
| GenBank mRNA | AK225967.1 | - | 8 | 1616 | ||
| BC007506.2 | - | 6 | 1776 | 3028787 | ||
| U68723.1 | - | 7 | 1826 | |||

FOXN3
Genomic Map