siRNA Details
| Gene Symbol: EPHA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005232.4 | NP_005223.4 | 7 | 1473 | ||
| XM_006715880.3 | XP_006715943.1 | 6 | 1101 | |||
| GenBank mRNA | AK290351.1 | - | 7 | 1437 | ||
| AK313263.1 | - | 7 | 1446 | |||
| BC130291.1 | - | 7 | 1457 | 40146322 | ||
| BC144156.1 | - | 6 | 1609 | |||
| CD653381.1 | - | 2 | 362 | |||
| EU826605.1 | - | 7 | 1414 | |||
| M18391.1 | - | 7 | 1479 | |||
| Z27409.1 | - | 3 | 531 | |||

EPHA1
Genomic Map