siRNA Details
| Gene Symbol: MS4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031666.1 | NP_001026836.1 | 3 | 305 | ||
| NM_001031809.1 | NP_001026979.1 | 4 | 476 | |||
| NM_006138.4 | NP_006129.4 | 5 | 614 | |||
| XM_011545363.2 | XP_011543665.1 | 4 | 361 | |||
| GenBank mRNA | AY095480.1 | - | 3 | 348 | ||
| BC008487.1 | - | 5 | 559 | 4102067 | ||
| BE564041.1 | - | 3 | 326 | |||
| BF210971.1 | - | 348 | ||||
| L35848.1 | - | 5 | 584 | |||

MS4A3
Genomic Map