siRNA Details
| Gene Symbol: MS4A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031666.1 | NP_001026836.1 | 5 | 565 | ||
| NM_001031809.1 | NP_001026979.1 | 6 | 736 | |||
| NM_006138.4 | NP_006129.4 | 7 | 874 | |||
| XM_011545363.2 | XP_011543665.1 | 6 | 621 | |||
| GenBank mRNA | BC008487.1 | - | 7 | 819 | 4102067 | |
| L35848.1 | - | 7 | 852 | |||

MS4A3
Genomic Map