siRNA Details
| Gene Symbol: LRBA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199282.2 | NP_001186211.2 | 3 | 655 | ||
| NM_006726.4 | NP_006717.2 | 3 | 614 | |||
| XM_005263372.3 | XP_005263429.1 | 3 | 800 | |||
| XM_005263373.2 | XP_005263430.1 | 3 | 842 | |||
| XM_005263374.3 | XP_005263431.1 | 3 | 799 | |||
| XM_005263375.3 | XP_005263432.1 | 3 | 799 | |||
| XM_011532434.2 | XP_011530736.1 | 3 | 799 | |||
| XM_017008872.1 | XP_016864361.1 | 3 | 842 | |||
| GenBank mRNA | AF216648.2 | - | 3 | 614 | ||
| AF467287.1 | - | 3 | 655 | |||
| AK098552.1 | - | 2 | 228 | |||

LRBA
Genomic Map