siRNA Details
| Gene Symbol: DIAPH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_006729.4 | NP_006720.1 | 16 | 2253 | ||
| NM_007309.3 | NP_009293.1 | 16 | 2253 | |||
| GenBank mRNA | AK002029.1 | - | 4 | 525 | ||
| AK291272.1 | - | 16 | 2135 | |||
| BC117414.1 | - | 17 | 1920 | 40125965 | ||
| BC143838.1 | - | 17 | 1920 | 9052353 | ||
| Y15908.1 | - | 16 | 2207 | |||
| Y15909.1 | - | 16 | 2207 | |||

DIAPH2
Genomic Map