siRNA Details
| Gene Symbol: EVC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001306090.1 | NP_001293019.1 | 21 | 3158 | ||
| NM_153717.2 | NP_714928.1 | 21 | 3161 | |||
| XM_006713865.3 | XP_006713928.1 | 20 | 3046 | |||
| XM_006713866.3 | XP_006713929.1 | 20 | 3043 | |||
| GenBank mRNA | AF216184.1 | - | 21 | 3161 | ||
| AF216185.1 | - | 21 | 3161 | |||

EVC
Genomic Map