siRNA Details
| Gene Symbol: BRINP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014618.2 | NP_055433.2 | 4 | 1003 | ||
| GenBank mRNA | AF027734.1 | - | 4 | 965 | ||
| BC021560.1 | - | 4 | 967 | |||
| BC065196.1 | - | 4 | 956 | 5245480 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BRINP1| Gene Symbol: BRINP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_014618.2 | NP_055433.2 | 4 | 1003 | ||
| GenBank mRNA | AF027734.1 | - | 4 | 965 | ||
| BC021560.1 | - | 4 | 967 | |||
| BC065196.1 | - | 4 | 956 | 5245480 | ||