siRNA Details
| Gene Symbol: CENPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016343.3 | NP_057427.3 | 6 | 935 | ||
| XM_011509082.2 | XP_011507384.1 | 6 | 943 | |||
| XM_017000086.1 | XP_016855575.1 | 6 | 922 | |||
| GenBank mRNA | AK307690.1 | - | 6 | 918 | ||
| U19769.1 | - | 931 | ||||
| U30872.1 | - | 6 | 833 | |||

CENPF
Genomic Map