siRNA Details
| Gene Symbol: CRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005206.4 | NP_005197.3 | 3 | 1287 | ||
| NM_016823.3 | NP_058431.2 | 3 | 1457 | |||
| GenBank mRNA | AK291060.1 | - | 3 | 1252 | ||
| BC001718.1 | - | 3 | 1409 | 3353237 | ||
| BC008506.1 | - | 3 | 1440 | 3452137 | ||
| BC009837.1 | - | 3 | 1253 | 4040536 | ||
| D10656.1 | - | 3 | 1414 | |||

CRK
Genomic Map