siRNA Details
| Gene Symbol: FANCD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018115.2 | NP_001018125.1 | 9 | 722 | ||
| NM_001319984.1 | NP_001306913.1 | 9 | 794 | |||
| NM_033084.4 | NP_149075.2 | 9 | 722 | |||
| GenBank mRNA | AF230336.1 | - | 9 | 665 | ||
| AF340183.1 | - | 9 | 665 | |||
| AK022613.1 | - | 2 | 124 | |||
| AK307512.1 | - | 10 | 695 | |||
| BC013582.2 | - | 9 | 653 | |||

FANCD2
Genomic Map