siRNA Details
| Gene Symbol: FGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042729.1 | NP_001036194.1 | 4 | 520 | ||
| NM_001042747.1 | NP_001036212.1 | 4 | 579 | |||
| NM_005248.2 | NP_005239.1 | 4 | 536 | |||
| XM_006710452.2 | XP_006710515.1 | 5 | 587 | |||
| XM_011541010.1 | XP_011539312.1 | 4 | 501 | |||
| XM_011541012.1 | XP_011539314.1 | 4 | 536 | |||
| XM_011541013.2 | XP_011539315.1 | 4 | 536 | |||
| GenBank mRNA | AK130188.1 | - | 4 | 498 | ||
| AK301078.1 | - | 4 | 492 | |||
| BC064382.1 | - | 4 | 449 | 5734966 | ||
| DA139476.1 | - | 4 | 536 | |||
| DA952771.1 | - | 4 | 475 | |||
| DA960178.1 | - | 4 | 503 | |||
| M19722.1 | - | 2 | 455 | |||
| M27454.1 | - | 4 | 461 | |||

FGR
Genomic Map