siRNA Details
| Gene Symbol: CETN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297765.1 | NP_001284694.1 | 2, 3 | 286 | ||
| NM_001297768.1 | NP_001284697.1 | 2, 3 | 286 | |||
| NM_004365.3 | NP_004356.2 | 2, 3 | 286 | |||
| GenBank mRNA | BC005383.1 | - | 2, 3 | 256 | 3935674 | |
| BC093793.1 | - | 2, 3 | 222 | 7939638 | ||
| BC112040.1 | - | 2, 3 | 222 | 8327508 | ||
| BM553126.1 | - | 2, 3 | 266 | |||
| BT006688.1 | - | 2, 3 | 161 | |||
| BU569574.1 | - | 3, 4 | 235 | |||
| CB988525.1 | - | 2, 3 | 308 | |||
| CB998764.1 | - | 2, 3 | 289 | |||
| CR450346.1 | - | 2, 3 | 161 | |||
| Y12473.1 | - | 3, 4 | 161 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_007684, Y12474, AK004532, AK017505, BC002162, and others.

CETN3
Genomic Map