siRNA Details
| Gene Symbol: C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000587.2 | NP_000578.2 | 18 | 3554 | ||
| GenBank mRNA | BC063851.1 | - | 18 | 3535 | 6184213 | |
| J03507.1 | - | 17 | 3438 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7| Gene Symbol: C7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000587.2 | NP_000578.2 | 18 | 3554 | ||
| GenBank mRNA | BC063851.1 | - | 18 | 3535 | 6184213 | |
| J03507.1 | - | 17 | 3438 | |||