siRNA Details
| Gene Symbol: ADRA2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000683.3 | NP_000674.2 | 1 | 1872 | ||
| GenBank mRNA | BC142625.1 | - | 1 | 2043 | 40147492 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ADRA2C| Gene Symbol: ADRA2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000683.3 | NP_000674.2 | 1 | 1872 | ||
| GenBank mRNA | BC142625.1 | - | 1 | 2043 | 40147492 | |
Set Membership: |
|