siRNA Details
| Gene Symbol: C4BPA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000715.3 | NP_000706.1 | 11 | 1680 | ||
| XM_005273251.1 | XP_005273308.1 | 11 | 1636 | |||
| XM_005273252.4 | XP_005273309.1 | 11 | 1812 | |||
| GenBank mRNA | AK303793.1 | - | 12 | 1491 | ||
| AK303798.1 | - | 11 | 1630 | |||
| AK313164.1 | - | 11 | 1626 | |||
| BC022312.1 | - | 11 | 1680 | 4688428 | ||
| M31452.1 | - | 11 | 1624 | |||
| X02865.1 | - | 9 | 1249 | |||

C4BPA
Genomic Map