siRNA Details
| Gene Symbol: ACTN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001104.3 | NP_001095.2 | 17 | 2240 | ||
| NM_001258371.2 | NP_001245300.2 | 17 | 2388 | |||
| NR_047663.2 | 17 | 2240 | ||||
| GenBank mRNA | AK303044.1 | - | 17 | 2388 | ||
| AK313556.1 | - | 17 | 2178 | |||
| BC096323.2 | - | 11 | 1300 | |||
| BC099647.3 | - | 17 | 2161 | 40008220 | ||
| BC099648.1 | - | 7 | 909 | |||
| BC099649.3 | - | 17 | 2161 | 40008223 | ||
| CX787471.1 | - | 2 | 234 | |||
| M86407.1 | - | 17 | 2161 | |||

ACTN3
Genomic Map